Learn More
Abnova™ Human LASS1 Partial ORF (NP_067090.1, 301 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00010715-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site that is cleaved to produce a mature protein containing seven conserved cysteine residues. Members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in yeast suggest that the encoded protein is involved in aging. This protein is transcribed from a monocistronic mRNA as well as a bicistronic mRNA, which also encodes growth differentiation factor 1. [provided by RefSeq]
Sequence: YIVAFAAKVLTGQVHELKDLREYDTAEAQSLKPSKAEKPLRNGLVKDKRFSpezifikation
NP_067090.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.24kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YIVAFAAKVLTGQVHELKDLREYDTAEAQSLKPSKAEKPLRNGLVKDKRF | |
RUO | |
LASS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10715 | |
LASS1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CerS1/LAG1/MGC90349/UOG1 | |
LASS1 | |
Recombinant | |
wheat germ expression system |