Learn More
Abnova™ Human KREMEN2 Full-length ORF (NP_078783.1, 1 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00079412-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. The encoded protein forms a ternary membrane complex with DKK1 and the WNT receptor lipoprotein receptor-related protein 6 (LRP6), and induces rapid endocytosis and removal of LRP6 from the plasma membrane. It contains extracellular kringle, WSC, and CUB domains. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. [provided by RefSeq]
Sequence: MGTQALQGFLFLLFLPLLQPRGASAGSLHSPGLSECFQVNGADYRGHQNRTGPRGAGRPCLFWDQTQQHSYSSASDPHGRWGLGAHNFCRNPDGDVQPWCYVAETEEGIYWRYCDIPSCHMPGYLGCFVDSGAPPALSGPSGTSTKLTVQVCLRFCRMKGYQLAGVEAGYACFCGSESDLARGRLAPATDCDQICFGHPGQLCGGDGRLGVYEVSVGSCQGNWTAPQGVIYSPDFPDEYGPDRNCSWALGPPGAALELTFRLFELADPRDRLELRDAASGSLLRAFDGARPPPSGPLRLGTAALLLTFRSDARGHAQGFALTYRGLQDAAEDPEAPEGSAQTPAAPLDGANVSCSPRPGAPPAAIGGAVCWLREKGPRRWGLPGAPGEAGLCGTNSPEGWPCPAPPGTPRLRVLPRATGLSpezifikation
NP_078783.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRM2/MGC10791/MGC16709 | |
KREMEN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
79412 | |
KREMEN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGTQALQGFLFLLFLPLLQPRGASAGSLHSPGLSECFQVNGADYRGHQNRTGPRGAGRPCLFWDQTQQHSYSSASDPHGRWGLGAHNFCRNPDGDVQPWCYVAETEEGIYWRYCDIPSCHMPGYLGCFVDSGAPPALSGPSGTSTKLTVQVCLRFCRMKGYQLAGVEAGYACFCGSESDLARGRLAPATDCDQICFGHPGQLCGGDGRLGVYEVSVGSCQGNWTAPQGVIYSPDFPDEYGPDRNCSWALGPPGAALELTFRLFELADPRDRLELRDAASGSLLRAFDGARPPPSGPLRLGTAALLLTFRSDARGHAQGFALTYRGLQDAAEDPEAPEGSAQTPAAPLDGANVSCSPRPGAPPAAIGGAVCWLREKGPRRWGLPGAPGEAGLCGTNSPEGWPCPAPPGTPRLRVLPRATGL | |
RUO | |
KREMEN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |