missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kell Antibody, R&D Systems™

Mouse Monoclonal Antibody
240.00 CHF - 654.00 CHF
Spezifikation
| Antigen | Kell |
|---|---|
| Klon | 195031 |
| Konzentration | LYOPH |
| Verdünnung | Western Blot 1 ug/mL |
| Anwendungen | Western Blot |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18706419
![]() |
R&D Systems
MAB1914-SP |
25 μg |
240.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18183251
![]() |
R&D Systems
MAB1914 |
500 μg |
654.00 CHF
500 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Kell Monoclonal specifically detects Kell in Human samples. It is validated for Western Blot.Spezifikation
| Kell | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P23276 | |
| 3792 | |
| Mouse myeloma cell line NS0-derived recombinant human Kell Asn68-Trp732 Accession # P23276 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Kell in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human ECE-2, recombinant human Neprilysin, or recombinant mouse Kell is observed. |
| 195031 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD238, CD238 antigen, EC 3.4.24.-, ECE3, KEL, Kell blood group, kell blood group antigen, kell blood group glycoprotein, Kell blood group, metalloendopeptidase, Kell blood group, metallo-endopeptidase | |
| KEL | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts