Learn More
Human Kell Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marke: R&D Systems MAB1914-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Kell Monoclonal specifically detects Kell in Human samples. It is validated for Western Blot.
Spezifikation
| Kell | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P23276 | |
| KEL | |
| Mouse myeloma cell line NS0-derived recombinant human Kell Asn68-Trp732 Accession # P23276 | |
| 25 μg | |
| Primary | |
| Detects human Kell in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human ECE-2, recombinant human Neprilysin, or recombinant mouse Kell is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 195031 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD238, CD238 antigen, EC 3.4.24.-, ECE3, KEL, Kell blood group, kell blood group antigen, kell blood group glycoprotein, Kell blood group, metalloendopeptidase, Kell blood group, metallo-endopeptidase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3792 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |