missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KEL Partial ORF (NP_000411.1, 633 a.a. - 732 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00003792-Q01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16130415

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases. [provided by RefSeq]

Sequence: LENAADVGGLAIALQAYSKRLLRHHGETVLPSLDLSPQQIFFRSYAQVMCRKPSPQDSHDTHSPPHLRVHGPLSSTPAFARYFRCARGALLNPSSRCQLW
Spezifikation

Spezifikation

NP_000411.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LENAADVGGLAIALQAYSKRLLRHHGETVLPSLDLSPQQIFFRSYAQVMCRKPSPQDSHDTHSPPHLRVHGPLSSTPAFARYFRCARGALLNPSSRCQLW
RUO
KEL
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3792
KEL (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD238/ECE3
KEL
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote