missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ITGAX Partial ORF (NP_000878, 114 a.a. - 223 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00003687-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16130295

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 25-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles. [provided by RefSeq]

Sequence: HECGRNMYLTGLCFLLGPTQLTQRLPVSRQECPRQEQDIVFLIDGSGSISSRNFATMMNFVRAVISQFQRPSTQFSLMQFSNKFQTHFTFEEFRRSSNPLSLLASVHQLQ
Spezifikation

Spezifikation

NP_000878
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HECGRNMYLTGLCFLLGPTQLTQRLPVSRQECPRQEQDIVFLIDGSGSISSRNFATMMNFVRAVISQFQRPSTQFSLMQFSNKFQTHFTFEEFRRSSNPLSLLASVHQLQ
RUO
ITGAX
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3687
ITGAX (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD11C/SLEB6
ITGAX
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote