missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-1 beta, Recombinant Protein
E. coli Recombinant Protein
Marke: Invitrogen™ A42508
Additional Details : Gewicht : 0.02180kg
Beschreibung
Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.Spezifikation
P01584 | |
protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human IL-1 beta | |
E. coli | |
RUO | |
IL1B | |
Unconjugated | |
Lyophilized |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
3553 | |
5 μg | |
-20°C | |
catabolin; cytokine; il 1 b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; Il-1 b; IL1B1; IL1beta; IL-1 beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1 beta-like; Interleukin-1 b; interleukin-beta; LOC100052414; lymphocyte proliferation-potentiating factor; preinterleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta | |
Human | |
Recombinant | |
≥ 90% |