Learn More
Abnova™ Human ID4 Full-length ORF (NP_001537.1, 1 a.a. - 161 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00003400-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Transcription factors containing a basic helix-loop-helix (bHLH) motif regulate expression of tissue-specific genes in a number of mammalian and insect systems. DNA-binding activity of the bHLH proteins is dependent on formation of homo- and/or heterodimers. Dominant-negative HLH proteins encoded by Id-related genes, such as ID4, also contain the HLH-dimerization domain but lack the DNA-binding basic domain. Consequently, Id proteins inhibit binding to DNA and transcriptional transactivation by heterodimerization with bHLH proteins (Pagliuca et al., 1995 [PubMed 7665172]).[supplied by OMIM]
Sequence: MKAVSPVRPSGRKAPSGCGGGELALRCLAEHGHSLGGSAAAAAAAAAARCKAAEAAADEPALCLQCDMNDCYSRLRRLVPTIPPNKKVSKVEILQHVIDYILDLQLALETHPALLRQPPPPAPPHHPAGTCPAAPPRTPLTALNTDPAGAVNKQGDSILCRSpezifikation
NP_001537.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IDB4/bHLHb27 | |
ID4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3400 | |
ID4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKAVSPVRPSGRKAPSGCGGGELALRCLAEHGHSLGGSAAAAAAAAAARCKAAEAAADEPALCLQCDMNDCYSRLRRLVPTIPPNKKVSKVEILQHVIDYILDLQLALETHPALLRQPPPPAPPHHPAGTCPAAPPRTPLTALNTDPAGAVNKQGDSILCR | |
RUO | |
ID4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |