Learn More
Abnova™ Human HSD3B7 Partial ORF (NP_079469, 101 a.a. - 209 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00080270-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes an enzyme which is involved in the initial stages of the synthesis of bile acids from cholesterol and a member of the short-chain dehydrogenase/reductase superfamily. The encoded protein is a membrane-associated endoplasmic reticulum protein which is active against 7-alpha hydrosylated sterol substrates. Mutations in this gene are associated with a congenital bile acid synthesis defect which leads to neonatal cholestasis, a form of progressive liver disease. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: IHEVNVQGTRNVIEACVQTGTRFLVYTSSMEVVGPNTKGHPFYRGNEDTPYEAVHRHPYPCSKALAEWLVLEANGRKVRGGLPLVTCALRPTGIYGEGHQIMRDFYRQGSpezifikation
NP_079469 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IHEVNVQGTRNVIEACVQTGTRFLVYTSSMEVVGPNTKGHPFYRGNEDTPYEAVHRHPYPCSKALAEWLVLEANGRKVRGGLPLVTCALRPTGIYGEGHQIMRDFYRQG | |
RUO | |
HSD3B7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
80270 | |
HSD3B7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PFIC4/SDR11E3 | |
HSD3B7 | |
Recombinant | |
wheat germ expression system |