missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HRB Full-length ORF (NP_004495.2, 1 a.a. - 562 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00003267-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16129037

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab:
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The protein encoded by this gene is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. The encoded protein binds the activation domain of the human immunodeficiency virus Rev protein when Rev is assembled onto its RNA target, and is required for the nuclear export of Rev-directed RNAs. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MAASAKRKQEEKHLKMLRDMTGLPHNRKCFDCDQRGPTYVNMTVGSFVCTSCSGSLRGLNPPHRVKSISMTTFTQQEIEFLQKHGNEVCKQIWLGLFDDRSSAIPDFRDPQKVKEFLQEKYEKKRWYVPPEQAKVVASVHASISGSSASSTSSTPEVKPLKSLLGDSAPTLHLNKGTPSQSPVVGRSQGQQQEKKQFDLLSDLGSDIFAAPAPQSTATANFANFAHFNSHAAQNSANADFANFDAFGQSSGSSNFGGFPTASHSPFQPQTTGGSAASVNANFAHFDNFPKSSSADFGTFNTSQSHQTASAVSKVSTNKAGLQTADKYAALANLDNIFSAGQGGDQGSGFGTTGKAPVGSVVSVPSQSSASSDKYAALAELDSVFSSAATSSNAYTSTSNASSNVFGTVPVVASAQTQPASSSVPAPFGATPSTNPFVAAAGPSVASSTNPFQTNARGATAATFGTASMSMPTGFGTPAPYSLPTSFSGSFQQPAFPAQAAFPQQTAFSQQPNGAGFAAFGQTKPVVTPFGQVAAAGVSSNPFMTGAPTGQFPTGSSSTNPFL
Spezifikation

Spezifikation

NP_004495.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
AGFG1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
3267
HRB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686I15205/HRB/MGC116938/MGC116940/RAB/RIP
AGFG1
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote