missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human HRB Full-length ORF (NP_004495.2, 1 a.a. - 562 a.a.) Recombinant Protein with GST-tag at N-terminal Artikelnummer.: 16119037
Verfügbar ab: 13-08-2025
Nur noch null auf Lager
Zum Warenkorb hinzufügen

Abnova™ Human HRB Full-length ORF (NP_004495.2, 1 a.a. - 562 a.a.) Recombinant Protein with GST-tag at N-terminal

Artikelnummer. p-7094639
10 ug, 10 Mikrogramm
Click to view available options
Menge:
10 ug
25 ug
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16119037

Marke: Abnova™ H00003267P01.10ug

Nur noch null auf Lager
Verfügbar ab: 13-08-2025
Zum Warenkorb hinzufügen
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. The encoded protein binds the activation domain of the human immunodeficiency virus Rev protein when Rev is assembled onto its RNA target, and is required for the nuclear export of Rev-directed RNAs. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MAASAKRKQEEKHLKMLRDMTGLPHNRKCFDCDQRGPTYVNMTVGSFVCTSCSGSLRGLNPPHRVKSISMTTFTQQEIEFLQKHGNEVCKQIWLGLFDDRSSAIPDFRDPQKVKEFLQEKYEKKRWYVPPEQAKVVASVHASISGSSASSTSSTPEVKPLKSLLGDSAPTLHLNKGTPSQSPVVGRSQGQQQEKKQFDLLSDLGSDIFAAPAPQSTATANFANFAHFNSHAAQNSANADFANFDAFGQSSGSSNFGGFPTASHSPFQPQTTGGSAASVNANFAHFDNFPKSSSADFGTFNTSQSHQTASAVSKVSTNKAGLQTADKYAALANLDNIFSAGQGGDQGSGFGTTGKAPVGSVVSVPSQSSASSDKYAALAELDSVFSSAATSSNAYTSTSNASSNVFGTVPVVASAQTQPASSSVPAPFGATPSTNPFVAAAGPSVASSTNPFQTNARGATAATFGTASMSMPTGFGTPAPYSLPTSFSGSFQQPAFPAQAAFPQQTAFSQQPNGAGFAAFGQTKPVVTPFGQVAAAGVSSNPFMTGAPTGQFPTGSSSTNPFL

Spezifikation

Zugriffsnummer NP_004495.2
Zur Verwendung mit (Anwendung) Antibody Production, Protein Array, ELISA, Western Blot
Zusammensetzung 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gen-ID (Entrez) 3267
Molekulargewicht 84.7kDa
Name HRB (Human) Recombinant Protein (P01)
Reinigungsverfahren Glutathione Sepharose 4 Fast Flow
Qualitätskontrollen 12.5% SDS-PAGE Stained with Coomassie Blue.
Menge 10 ug
Lagerungsbedingungen Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Kennzeichnung RUO
Gen-Alias DKFZp686I15205/HRB/MGC116938/MGC116940/RAB/RIP
Gebräuchliche Bezeichnung AGFG1
Gensymbol AGFG1
Spezies Wheat Germ (in vitro)
Rekombinant Recombinant
Proteinmarkierung GST
Expressionssystem wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human HRB Full-length ORF (NP_004495.2, 1 a.a. - 562 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.