Learn More
Abnova™ Human HRB Full-length ORF (NP_004495.2, 1 a.a. - 562 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00003267-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. The encoded protein binds the activation domain of the human immunodeficiency virus Rev protein when Rev is assembled onto its RNA target, and is required for the nuclear export of Rev-directed RNAs. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MAASAKRKQEEKHLKMLRDMTGLPHNRKCFDCDQRGPTYVNMTVGSFVCTSCSGSLRGLNPPHRVKSISMTTFTQQEIEFLQKHGNEVCKQIWLGLFDDRSSAIPDFRDPQKVKEFLQEKYEKKRWYVPPEQAKVVASVHASISGSSASSTSSTPEVKPLKSLLGDSAPTLHLNKGTPSQSPVVGRSQGQQQEKKQFDLLSDLGSDIFAAPAPQSTATANFANFAHFNSHAAQNSANADFANFDAFGQSSGSSNFGGFPTASHSPFQPQTTGGSAASVNANFAHFDNFPKSSSADFGTFNTSQSHQTASAVSKVSTNKAGLQTADKYAALANLDNIFSAGQGGDQGSGFGTTGKAPVGSVVSVPSQSSASSDKYAALAELDSVFSSAATSSNAYTSTSNASSNVFGTVPVVASAQTQPASSSVPAPFGATPSTNPFVAAAGPSVASSTNPFQTNARGATAATFGTASMSMPTGFGTPAPYSLPTSFSGSFQQPAFPAQAAFPQQTAFSQQPNGAGFAAFGQTKPVVTPFGQVAAAGVSSNPFMTGAPTGQFPTGSSSTNPFLSpezifikation
NP_004495.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
84.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AGFG1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
3267 | |
HRB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686I15205/HRB/MGC116938/MGC116940/RAB/RIP | |
AGFG1 | |
Recombinant | |
wheat germ expression system |