Learn More
Abnova™ Human GUCA1A Partial ORF (NP_000400, 1 a.a. - 93 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002978-Q01.25ug
Beschreibung
This gene plays a role in the recovery of retinal photoreceptors from photobleaching. In the recovery phase, the phototransduction messeneger cGMP is replenished by retinal guanylyl cyclase-1 (GC1). GC1 is activated by decreasing Ca(2+) concentrations following photobleaching. The protein encoded by this gene, guanylyl cyclase activating protein 1 (GCAP1), mediates the sensitivity of GC1 to Ca(2+) concentrations. GCAP1 promotes activity of GC1 at low Ca(2+) concentrations and inhibits GC1 activity at high Ca(2+) concentrations. Mutations in this gene cause autosomal dominant cone dystrophy (COD3); a disease characterized by reduced visual acuity associated with progressive loss of color vision. Mutations in this gene prohibit the inactivation of RetGC1 at high Ca(2+) concentrations; causing the constitutive activation of RetGC1 and, presumably, increased cell death. This gene is expressed in retina and spermatagonia. [provided by RefSeq]
Sequence: MGNVMEGKSVEELSSTECHQWYKKFMTECPSGQLTLYEFRQFFGLKNLSPSASQYVEQMFETFDFNKDGYIDFMEYVAALSLVLKGKVEQKLRSpezifikation
NP_000400 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.97kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGNVMEGKSVEELSSTECHQWYKKFMTECPSGQLTLYEFRQFFGLKNLSPSASQYVEQMFETFDFNKDGYIDFMEYVAALSLVLKGKVEQKLR | |
RUO | |
GUCA1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2978 | |
GUCA1A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COD3/GCAP/GCAP1/GUCA/GUCA1 | |
GUCA1A | |
Recombinant | |
wheat germ expression system |