Learn More
Abnova™ Human GRK5 Partial ORF (AAH64506, 51 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002869-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes (PMNs). [provided by RefSeq]
Sequence: RDYCSLCDKQPIGRLLFRQFCETRPGLECYIQFLDSVAEYEVTPDEKLGEKGKEIMTKYLTPKSPVFIAQVGQDLVSQTEEKLLQKPCKELFSACAQSVHSpezifikation
AAH64506 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RDYCSLCDKQPIGRLLFRQFCETRPGLECYIQFLDSVAEYEVTPDEKLGEKGKEIMTKYLTPKSPVFIAQVGQDLVSQTEEKLLQKPCKELFSACAQSVH | |
RUO | |
GRK5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2869 | |
GRK5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPRK5 | |
GRK5 | |
Recombinant | |
wheat germ expression system |