missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GRK5 Partial ORF (AAH64506, 51 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | AAH64506 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 2869 |
Molekulargewicht | 36.63kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16128864
|
Abnova™
H00002869-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16118864
|
Abnova™
H00002869-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes (PMNs). [provided by RefSeq]
Sequence: RDYCSLCDKQPIGRLLFRQFCETRPGLECYIQFLDSVAEYEVTPDEKLGEKGKEIMTKYLTPKSPVFIAQVGQDLVSQTEEKLLQKPCKELFSACAQSVHSpezifikation
AAH64506 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPRK5 | |
GRK5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2869 | |
GRK5 (Human) Recombinant Protein (Q01) | |
RDYCSLCDKQPIGRLLFRQFCETRPGLECYIQFLDSVAEYEVTPDEKLGEKGKEIMTKYLTPKSPVFIAQVGQDLVSQTEEKLLQKPCKELFSACAQSVH | |
RUO | |
GRK5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |