missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GAN Partial ORF (NP_071324, 534 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_071324 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 8139 |
Molekulargewicht | 32.78kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16128255
|
Abnova™
H00008139-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 06-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16118255
|
Abnova™
H00008139-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 06-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of the cytoskeletal BTB/kelch (Broad-Complex, Tramtrack and Bric a brac) repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy (GAN). [provided by RefSeq]
Sequence: DLDTGTNYDYVREFKRSTGTWHHTKPLLPSDLRRTGCAALRIANCKLFRLQLQQGLFRIRVHSPSpezifikation
NP_071324 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.78kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ38059/GAN1/KLHL16 | |
GAN | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8139 | |
GAN (Human) Recombinant Protein (Q01) | |
DLDTGTNYDYVREFKRSTGTWHHTKPLLPSDLRRTGCAALRIANCKLFRLQLQQGLFRIRVHSP | |
RUO | |
GAN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |