Learn More
Abnova™ Human GAN Partial ORF (NP_071324, 534 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008139-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the cytoskeletal BTB/kelch (Broad-Complex, Tramtrack and Bric a brac) repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy (GAN). [provided by RefSeq]
Sequence: DLDTGTNYDYVREFKRSTGTWHHTKPLLPSDLRRTGCAALRIANCKLFRLQLQQGLFRIRVHSPSpezifikation
NP_071324 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.78kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DLDTGTNYDYVREFKRSTGTWHHTKPLLPSDLRRTGCAALRIANCKLFRLQLQQGLFRIRVHSP | |
RUO | |
GAN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8139 | |
GAN (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ38059/GAN1/KLHL16 | |
GAN | |
Recombinant | |
wheat germ expression system |