missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human FKBP4 / FKBP52 Protein, His,PA Tag
479.18 CHF - 2522.00 CHF
Spezifikation
Zugriffsnummer | AAH01786 |
---|---|
Molekulargewicht | 55.1 kDa |
Quelle | Human FKBP4, His,PA Tag (FK4-H52Q3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 459 (Accession # AAH01786). |
Immunogen | Met 1 - Ala 459 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15816767
|
ACROBiosystems
FK4-H52Q3-500UG |
500 μg |
2522.00 CHF
500 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15826767
|
ACROBiosystems
FK4-H52Q3-50UG |
50 μg |
479.18 CHF
50 Mikrogramm |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Description
Peptidyl-prolyl cis-trans isomerase FKBP4 (PPIase FKBP4) is also known as immunophilin FKBP52. FKBP4 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP4 has high structural and functional similarity to FK506-binding protein 1A (FKBP1A), but unlike FKBP1A, this protein does not have immunosuppressant activity when complexed with FK506. FKBP4 interacts with interferon regulatory factor-4 and plays an important role in immunoregulatory gene expression in B and T lymphocytes.
This protein carries a polyhistidine tag at the C-terminus, followed by a PA tag, and has a calculated MW of 55.1 kDa. The reducing (R) protein migrates as 60 kDa in SDS-PAGE.Spécification
AAH01786 | |
Human FKBP4, His,PA Tag (FK4-H52Q3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 459 (Accession # AAH01786). | |
-20°C | |
FKBP4 | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
55.1 kDa | |
Met 1 - Ala 459 | |
FKBP4,FKBP52 | |
Unconjugated | |
His Tag and PA Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |