Learn More
ACROBiosystems Human FKBP4 / FKBP52 Protein, His,PA Tag
Marke:  ACROBiosystems FK4-H52Q3-50UG
Additional Details : Gewicht : 0.00001kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15826767
  • 479.18 CHF / 50 Mikrogramm
Verfügbar ab: 15-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Peptidyl-prolyl cis-trans isomerase FKBP4 (PPIase FKBP4) is also known as immunophilin FKBP52. FKBP4 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP4 has high structural and functional similarity to FK506-binding protein 1A (FKBP1A), but unlike FKBP1A, this protein does not have immunosuppressant activity when complexed with FK506. FKBP4 interacts with interferon regulatory factor-4 and plays an important role in immunoregulatory gene expression in B and T lymphocytes.
This protein carries a polyhistidine tag at the C-terminus, followed by a PA tag, and has a calculated MW of 55.1 kDa. The reducing (R) protein migrates as 60 kDa in SDS-PAGE.
Spezifikation
Spezifikation
AAH01786
50 μg
Met 1 - Ala 459
FKBP4,FKBP52
Unconjugated
His Tag and PA Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
55.1 kDa
Human FKBP4, His,PA Tag (FK4-H52Q3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 459 (Accession # AAH01786).
-20°C
FKBP4
Human
50 mM Tris, 100 mM Glycine, pH7.5
HEK293
Other Recombinant Proteins