missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FGF-10, Recombinant Protein
E. coli Recombinant Protein
Marke: Invitrogen™ A42544
Additional Details : Gewicht : 0.01810kg
Beschreibung
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.Spezifikation
O15520 | |
protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human FGF-10 | |
E. coli | |
RUO | |
FGF10 | |
Unconjugated | |
Lyophilized |
Control, ELISA, Immunohistochemistry, Western Blot | |
2255 | |
5 x 5 μg | |
-20°C | |
AEY17; BB213776; FGF; FGF10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria | |
Human | |
Recombinant | |
≥ 90% |