missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human FAM171B Protein, His Tag
Marke: ACROBiosystems FAB-H5220-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Family with sequence similarity 171, member B (FAM171B) is also known as KIAA1946, NPD019, which is a single-pass type I membrane protein, belongs to the FAM171 family.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.8 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH60872 | |
1 mg | |
Glu 33 - Thr 355 | |
FAM171B,KIAA1946,NPD019 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
36.8 kDa | |
Human FAM171B, His Tag (FAB-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Thr 355 (Accession # AAH60872). | |
-20°C | |
FAM171B | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |