missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ESPL1 Partial ORF (NP_036423, 586 a.a. - 701 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00009700-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16151176

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 12-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Stable cohesion between sister chromatids before anaphase and their timely separation during anaphase are critical for chromosome inheritance. In vertebrates, sister chromatid cohesion is released in 2 steps via distinct mechanisms. The first step involves phosphorylation of STAG1 (MIM 604358) or STAG2 (MIM 604359) in the cohesin complex. The second step involves cleavage of the cohesin subunit SCC1 (RAD21; MIM 606462) by ESPL1, or separase, which initiates the final separation of sister chromatids (Sun et al., 2009 [PubMed 19345191]).[supplied by OMIM]

Sequence: REELQAYKAVRADTGQERFNIICDLLELSPEETPAGAWARATHLVELAQVLCYHDFTQQTNCSALDAIREALQLLDSVRPEAQARDQLLDDKAQALLWLYICTLEAKIQEGIERDR
Spezifikation

Spezifikation

NP_036423
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.5kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
REELQAYKAVRADTGQERFNIICDLLELSPEETPAGAWARATHLVELAQVLCYHDFTQQTNCSALDAIREALQLLDSVRPEAQARDQLLDDKAQALLWLYICTLEAKIQEGIERDR
RUO
ESPL1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9700
ESPL1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ESP1/FLJ46492/KIAA0165/SEPARASE/SEPARIN
ESPL1
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote