Learn More
Abnova™ Human DOK6 Full-length ORF (NP_689934.2, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00220164-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
DOK6 is a member of the DOK (see DOK1; MIM 602919) family of intracellular adaptors that play a role in the RET (MIM 164761) signaling cascade (Crowder et al., 2004 [PubMed 15286081]).[supplied by OMIM]
Sequence: MASNFNDIVKQGYVKIRSRKLGIFRRCWLVFKKASSKGPRRLEKFPDEKAAYFRNFHKVTELHNIKNITRLPRETKKHAVAIIFHDETSKTFACESELEAEEWCKHLCMECLGTRLNDISLGEPDLLAAGVQREQNERFNVYLMPTPNLDIYGECTMQITHENIYLWDIHNAKVKLVMWPLSSLRRYGRDSTWFTFESGRMCDTGEGLFTFQTREGEMIYQKVHSATLAIAEQHERLMLEMEQKARLQTSLTEPMTLSKSISLPRSAYWHHITRQNSVGEIYSLQGHGFGSSKMSRAQTFPSYAPEQSEEAQQPLSRSSSYGFSYSSSLIQSpezifikation
NP_689934.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DOK5L/HsT3226/MGC20785 | |
DOK6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
220164 | |
DOK6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASNFNDIVKQGYVKIRSRKLGIFRRCWLVFKKASSKGPRRLEKFPDEKAAYFRNFHKVTELHNIKNITRLPRETKKHAVAIIFHDETSKTFACESELEAEEWCKHLCMECLGTRLNDISLGEPDLLAAGVQREQNERFNVYLMPTPNLDIYGECTMQITHENIYLWDIHNAKVKLVMWPLSSLRRYGRDSTWFTFESGRMCDTGEGLFTFQTREGEMIYQKVHSATLAIAEQHERLMLEMEQKARLQTSLTEPMTLSKSISLPRSAYWHHITRQNSVGEIYSLQGHGFGSSKMSRAQTFPSYAPEQSEEAQQPLSRSSSYGFSYSSSLIQ | |
RUO | |
DOK6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |