missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DDO Partial ORF (NP_003640, 270 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00008528-Q01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16128735

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 13-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The protein encoded by this gene is a peroxisomal flavoprotein that catalyzes the oxidative deamination of D-aspartate and N-methyl D-aspartate. Flavin adenine dinucleotide or 6-hydroxyflavin adenine dinucleotide can serve as the cofactor in this reaction. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: WNLSPDAENSREILSRCCALEPSLHGACNIREKVGLRPYRPGVRLQTELLARDGQRLPVVHHYGHGSGGISVHWGTALEAARLVSECVHALRTPIPKSNL
Spezifikation

Spezifikation

NP_003640
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
WNLSPDAENSREILSRCCALEPSLHGACNIREKVGLRPYRPGVRLQTELLARDGQRLPVVHHYGHGSGGISVHWGTALEAARLVSECVHALRTPIPKSNL
RUO
DDO
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8528
DDO (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DASOX/DDO-1/DDO-2/FLJ45203
DDO
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote