missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CMTM6 Full-length ORF (NP_060271.1, 1 a.a. - 183 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00054918-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16120893

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 12-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq]

Sequence: MENGAVYSPTTEEDPGPARGPRSGLAAYFFMGRLPLLRRVLKGLQLLLSLLAFICEEVVSQCTLCGGLYFFEFVSCSAFLLSLLILIVYCTPFYERVDTTKVKSSDFYITLGTGCVFLLASIIFVSTHDRTSAEIAAIVFGFIASFMFLLDFITMLYEKRQESQLRKPENTTRAEALTEPLNA
Spezifikation

Spezifikation

NP_060271.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CKLFSF6/FLJ20396/PRO2219
CMTM6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54918
CMTM6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MENGAVYSPTTEEDPGPARGPRSGLAAYFFMGRLPLLRRVLKGLQLLLSLLAFICEEVVSQCTLCGGLYFFEFVSCSAFLLSLLILIVYCTPFYERVDTTKVKSSDFYITLGTGCVFLLASIIFVSTHDRTSAEIAAIVFGFIASFMFLLDFITMLYEKRQESQLRKPENTTRAEALTEPLNA
RUO
CMTM6
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote