missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CD6 Partial ORF (NP_006716, 308 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00000923-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16135804

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 11-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

CD6 is a monomeric 105- or 130-kD membrane glycoprotein that is involved in T-cell activation. The size difference between the 2 CD6 forms is due to differences in phosphorylation (Robinson et al., 1995 [PubMed 7589069]).[supplied by OMIM]

Sequence: CGTAVERPKGLPHSLSGRMYYSCNGEELTLSNCSWRFNNSNLCSQSLAARVLCSASRSLHNLSTPEVPASVQTVTIESSVTVKIENKESRELM
Spezifikation

Spezifikation

NP_006716
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.97kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CGTAVERPKGLPHSLSGRMYYSCNGEELTLSNCSWRFNNSNLCSQSLAARVLCSASRSLHNLSTPEVPASVQTVTIESSVTVKIENKESRELM
RUO
CD6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
923
CD6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ44171/TP120
CD6
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote