missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD14 Antibody, R&D Systems™

Goat Polyclonal Antibody
298.00 CHF - 569.00 CHF
Spezifikation
| Antigen | CD14 |
|---|---|
| Verdünnung | 0.5 μg/mL (Western Blot), 20 μg/mL (Simple Western), 5 to 25 μg/mL (Immunocytochemistry) |
| Anwendungen | Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18726285
![]() |
R&D Systems
AF3831-100 |
100 μg |
569.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18766295
![]() |
R&D Systems
AF3831-SP |
25 μg |
298.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
CD14 Polyclonal antibody specifically detects CD14 in Human samples. It is validated for Western Blot, Western Blot, ImmunocytochemistrySpezifikation
| CD14 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Goat | |
| Human | |
| P08571 | |
| 929 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
| 0.5 μg/mL (Western Blot), 20 μg/mL (Simple Western), 5 to 25 μg/mL (Immunocytochemistry) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Chinese Hamster Ovary cell line, CHO-derived human CD14 protein, Thr20-Cys352, Accession # P08571 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title