missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD14 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marke: R&D Systems AF3831-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD14 Polyclonal antibody specifically detects CD14 in Human samples. It is validated for Western Blot, Western Blot, Immunocytochemistry
Spezifikation
| CD14 | |
| Polyclonal | |
| 0.5 μg/mL (Western Blot), 20 μg/mL (Simple Western), 5 to 25 μg/mL (Immunocytochemistry) | |
| P08571 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 929 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Chinese Hamster Ovary cell line, CHO-derived human CD14 protein, Thr20-Cys352, Accession # P08571 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur