missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Cathepsin S / CTSS Protein, His Tag
auf Anfrage
Spezifikation
Zugriffsnummer | NP_004070.3 |
---|---|
Molekulargewicht | 24.9 kDa |
Quelle | Human Cathepsin S, His Tag (CTS-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 17 - Ile 331 (Accession # P25774-1). |
Immunogen | Gln 17 - Ile 331 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15826547
|
ACROBiosystems
CTS-H5224-1MG |
1 mg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
15836547
|
ACROBiosystems
CTS-H5224-50UG |
50 µg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Cathepsin S is also known as CTSS, which is a member of the peptidase C1 family. Cathepsin S is a lysosomal cysteine protease that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. Cathepsin S can function as an elastase over a broad pH range in alveolar macrophages. The catalytic activity of cathepsin S is similar to cathepsin L, but with much less activity on Z-Phe-Arg-|-NHMec, and more activity on the Z-Val-Val-Arg-|-Xaa compound.
This protein carries a polyhistidine tag at the C-terminus. This protein undergoes autocatalytic cleavage to release the propeptide and mature chain. The propeptide and mature chain are associated through non‐covalent interactions and with a calculated MSpezifikation
NP_004070.3 | |
Human Cathepsin S, His Tag (CTS-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 17 - Ile 331 (Accession # P25774-1). | |
-20°C | |
Cathepsin S | |
Human | |
50 mM MES, 100 mM NaCl, pH6.5 | |
HEK293 | |
Enzymes |
24.9 kDa | |
Gln 17 - Ile 331 | |
CTSS,Cathepsin S | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |