Learn More
ACROBiosystems Human Cathepsin S / CTSS Protein, His Tag
Marke: ACROBiosystems CTS-H5224-50UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Cathepsin S is also known as CTSS, which is a member of the peptidase C1 family. Cathepsin S is a lysosomal cysteine protease that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. Cathepsin S can function as an elastase over a broad pH range in alveolar macrophages. The catalytic activity of cathepsin S is similar to cathepsin L, but with much less activity on Z-Phe-Arg-|-NHMec, and more activity on the Z-Val-Val-Arg-|-Xaa compound.
This protein carries a polyhistidine tag at the C-terminus. This protein undergoes autocatalytic cleavage to release the propeptide and mature chain. The propeptide and mature chain are associated through non‐covalent interactions and with a calculated MSpezifikation
NP_004070.3 | |
50 µg | |
Gln 17 - Ile 331 | |
CTSS,Cathepsin S | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
24.9 kDa | |
Human Cathepsin S, His Tag (CTS-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 17 - Ile 331 (Accession # P25774-1). | |
-20°C | |
Cathepsin S | |
Human | |
50 mM MES, 100 mM NaCl, pH6.5 | |
HEK293 | |
Enzymes |