Learn More
Abnova™ Human ATPAF1 Partial ORF (NP_073582, 219 a.a. - 328 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00064756-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes an assembly factor for the F(1) component of the mitochondrial ATP synthase. This protein binds specifically to the F1 beta subunit and is thought to prevent this subunit from forming nonproductive homooligomers during enzyme assembly. Alternatively spliced transcript variants have been identified, but the biological validity of some of these variants has not been determined. [provided by RefSeq]
Sequence: LHFTALINIQTRGEAAASQLILYHYPELKEEKGIVLMTAEMDSTFLNVAEAQCIANQVQLFYATDRKETYGLVETFNLRPNEFKYMSVIAELEQSGLGAELKCAQNQNKTSpezifikation
NP_073582 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LHFTALINIQTRGEAAASQLILYHYPELKEEKGIVLMTAEMDSTFLNVAEAQCIANQVQLFYATDRKETYGLVETFNLRPNEFKYMSVIAELEQSGLGAELKCAQNQNKT | |
RUO | |
ATPAF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64756 | |
ATPAF1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP11/ATP11p/FLJ22351/MGC88060 | |
ATPAF1 | |
Recombinant | |
wheat germ expression system |