Learn More
Abnova™ Human ALDH8A1 Full-length ORF (NP_072090.1, 1 a.a. - 487 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00064577-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This protein belongs to the aldehyde dehydrogenases family of proteins. It plays a role in a pathway of 9-cis-retinoic acid biosynthesis in vivo. This enzyme converts 9-cis-retinal into the retinoid X receptor ligand 9-cis-retinoic acid, and has approximately 40-fold higher activity with 9-cis-retinal than with all-trans-retinal. Therefore, it is the first known aldehyde dehydrogenase to show a preference for 9-cis-retinal relative to all-trans-retinal. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MAGTNALLMLENFIDGKFLPCSSYIDSYDPSTGEVYCRVPNSGKDEIEAAVKAAREAFPSWSSRSPQERSRVLNQVADLLEQSLEEFAQAESKDQGKTLALARTMDIPRSVQNFRFFASSSLHHTSECTQMDHLGCMHYTVRAPVGVAGLISPWNLPLYLLTWKIAPAMAAGNTVIAKPSELTSVTAWMLCKLLDKAGVPPGVVNIVFGTGPRVGEALVSHPEVPLISFTGSQPTAERITQLSAPHCKKLSLELGGKNPAIIFEDANLDECIPATVRSSFANQGEICLCTSRIFVQKSIYSEFLKRFVEATRKWKVGIPSDPLVSIGALISKAHLEKVRSYVKRALAEGAQIWCGEGVDKLSLPARNQAGYFMLPTVITDIKDESCCMTEEIFGPVTCVVPFDSEEEVIERANNVKYGLAATVWSSNVGRVHRVAKKLQSGLVWTNCWLIRELNLPFGGMKSSGIGREGAKDSYDFFTEIKTITVKHSpezifikation
NP_072090.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ALDH8A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64577 | |
ALDH8A1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALDH12/DJ352A20.2/DKFZp779D2315/MGC138650 | |
ALDH8A1 | |
Recombinant | |
wheat germ expression system |