missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIPK4 Recombinant Human Protein
HIPK4 is a homeodomain-interacting protein kinase.
Marke: Invitrogen™ PV4176
Additional Details : Gewicht : 0.00900kg
Beschreibung
HIPK4 has been demonstrated to phosphorylate p53.
Spezifikation
NP_653286 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
HIPK4 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
96.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
DYRK Kinase Family | |
Recombinant | |
Baculovirus |