Learn More
Abeba™ High Original 5008 Shoes
Product Code. 15671768
Click to view available options
Size:
36
37
38
39
40
41
42
Unit Size:
1 pair
Product Code. 15671768
Brand: Abeba™ 5008/38
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 618 41 11 for assistance
View alternative products
ESD dissipative shoe includes PU sole with high slip resistance.
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather, perforated
  • White
  • Slip resistance PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
  • Narrow fit
  • Wedge heel 45mm
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color White
Material Leather
Size 38
Sole Features Slip-resistant
Sole Material Polyurethane
Style 5008