missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HHIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11654-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein similar to the mouse hedgehog-interacting protein, a regulatory component of the hedgehog signaling pathway. Members of the hedgehog family are evolutionarily conserved proteins which are involved in many fundamental processes in embryonic development, including anteroposterior patterns of limbs and regulation of left-right asymmetry.Spezifikation
HHIP | |
Polyclonal | |
Unconjugated | |
HHIP | |
hedgehog interacting protein, HHIP, HIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15245, 291936, 64399 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7TN16, Q96QV1 | |
Hhip | |
HHIP Fusion Protein Ag2241 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |