Learn More
HER2/ErbB2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18299-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody raised against a synthesized peptide corresponding to 1237-1255aa of human HER2 recognizes the 185- kDa full-length glycosylated form and other lower molecular-mass forms of HER2, including the truncated form.
This protein is a regulatory subunit of the condensin-2 complex, a complex which establishes mitotic chromosome architecture and is involved in physical rigidity of the chromatid axis.Spezifikation
HER2/ErbB2 | |
Polyclonal | |
Unconjugated | |
ERBB2 | |
CD340, ERBB2, HER 2, HER 2/neu, HER2, MLN 19, MLN19, NEU, NGL, p185, p185erbB2, Proto oncogene c ErbB 2, Proto oncogene Neu, TKR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2064 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04626 | |
ERBB2 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |