Learn More
Heidolph™ Hei-Mix L
Artikelnummer. 17303440 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH-Ausführung
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17303440 CH-Ausführung 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17303440 Lieferant Heidolph™ Lieferanten-Nr. 5050000004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
The Hei-Mix L was especially developed for applications with thermolabile substances.
  • Stirring up to 20 liters without heating
  • The Hei-Mix L was especially developed for applications with thermolabile substances. This is because this model has no heating, and the heat from the motor is also not transferred to the footprint.
  • The Hei-Mix L magnetic stirrer without heating function is ideal for applications up to 20 liters and can gently stir your solution in a speed range of 100 to 1400 rpm. In addition, the rotation speed ramps up slowly until it has reached the desired value. This prevents splashes, uneven starts, or jumps of the stirring rod.
  • The patented Kera-Disk™ footprint made of aluminum with a ceramic coating, is chemical-resistant, scratch-proof, and has a diameter of 145 mm. This unique size makes it suitable for use with many accessories and samples on a small footprint.
TRUSTED_SUSTAINABILITY
Spezifikation
Rührpositionen 1
Spannung 230 V
Steckertyp CH-Ausführung