missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HECTD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11487-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene transfers ubiquitin from an E2 ubiquitin-conjugating enzyme to targeted substrates, leading to the degradation of those substrates. The encoded protein has been shown to transfer ubiquitin to TRIOBP to facilitate cell cycle progression, and to STX8.Spezifikation
HECTD3 | |
Polyclonal | |
Unconjugated | |
HECTD3 | |
HECT domain containing 3, HECTD3, RP11 69J16.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
76608, 79654 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3U487, Q5T447 | |
Hectd3 | |
HECTD3 Fusion Protein Ag1998 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |