Learn More
HACL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15519-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
HACL1 gene ontology annotations related to this gene include lyase activity; magnesium ion binding; thiamine pyrophosphate binding.Spezifikation
| HACL1 | |
| Polyclonal | |
| Unconjugated | |
| HACL1 | |
| 2 HPCL, 2 hydroxyacyl CoA lyase 1, 2 hydroxyphytanoyl CoA lyase, HACL1, HPCL, HPCL2, PHYH2, Phytanoyl CoA 2 hydroxylase 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26061, 56794, 85255 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CHM7, Q9QXE0, Q9UJ83 | |
| Hacl1 | |
| HACL1 Fusion Protein Ag7830 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |