missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTSE1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21319-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is only expressed in the S and G2 phases of the cell cycle, where it colocalizes with cytoplasmic tubulin and microtubules. In response to DNA damage, the encoded protein accumulates in the nucleus and binds the tumor suppressor protein p53, shuttling it out of the nucleus and repressing its ability to induce apoptosis.Spezifikation
GTSE1 | |
Polyclonal | |
Unconjugated | |
GTSE1 | |
B99, G 2 and S phase expressed 1, GTSE 1, GTSE1, Protein B99 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51512 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NYZ3 | |
GTSE1 | |
GTSE1 Fusion Protein Ag13497 | |
150 μL | |
Primary | |
Human | |
Liquid |