missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTSE1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | GTSE1 |
---|---|
Konzentration | 0.3 mg/mL |
Anwendungen | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850925
|
Proteintech
21319-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 24-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840925
|
Proteintech
21319-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 24-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is only expressed in the S and G2 phases of the cell cycle, where it colocalizes with cytoplasmic tubulin and microtubules. In response to DNA damage, the encoded protein accumulates in the nucleus and binds the tumor suppressor protein p53, shuttling it out of the nucleus and repressing its ability to induce apoptosis.Spezifikation
GTSE1 | |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q9NYZ3 | |
51512 | |
GTSE1 Fusion Protein Ag13497 | |
Primary | |
-20°C | |
GTSE1 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B99, G 2 and S phase expressed 1, GTSE 1, GTSE1, Protein B99 homolog | |
GTSE1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |