missing translation for 'onlineSavingsMsg'
Learn More

GTF2A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 21381-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16850985

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 22-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A) and several general initiation factors. One of these factors is TFIIA, which when purified from HeLa extracts consists of 35-, 19-, and 12-kD subunits.
Spezifikation

Spezifikation

GTF2A1
Polyclonal
Unconjugated
GTF2A1
GTF2A1, TF2A1, TFIIA, TFIIA 42, TFIIA p19 subunit, TFIIAL
Rabbit
Antigen Affinity Chromatography
RUO
2957, 83602, 83830
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08949, P52655, Q99PM3
GTF2A1
GTF2A1 Fusion Protein Ag15994
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote