missing translation for 'onlineSavingsMsg'
Learn More

GSDMD Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 20770-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16840335

  • 351.68 CHF / 150 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

FOXN3 is a member of the forkhead/winged helixtranscription factor family. Checkpoints are eukaryotic DNA damage-inducible cell cycle arrests at G1 and G2. Check point suppressor 1 suppresses multiple yeast checkpoint mutationsincluding mec1, rad9, rad53 and dun1 by activating aMEC1-independent checkpoint pathway. Alternative splicing isobserved at the locus, resulting in distinct isoforms.
Spezifikation

Spezifikation

GSDMD
Polyclonal
Unconjugated
GSDMD
DF5L, DFNA5L, FKSG10, gasdermin D, GSDMD, GSDMDC1
Rabbit
Antigen Affinity Chromatography
RUO
79792
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P57764
GSDMD
GSDMD Fusion Protein Ag14757
150 μL
Primary
Human
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote