missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRASP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20020-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody is specific to GPRASP1.
Modulates lysosomal sorting and functional down-regulation of a variety of G-protein coupled receptors. Targets receptors for degradation in lysosomes via its interaction with BECN2.Spezifikation
GPRASP1 | |
Polyclonal | |
Unconjugated | |
GPRASP1 | |
GASP, GASP 1, GASP1, GPRASP1, KIAA0443 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67298, 9737 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5JY77, Q5U4C1 | |
Gprasp1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |