missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR142 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18820-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody recognizes the 51- kDa GRP142.
GPR142 is a member of the rhodopsin family of G protein-coupled receptors.Spezifikation
GPR142 | |
Polyclonal | |
Unconjugated | |
GPR142 | |
G protein coupled receptor 142, GPR142, PGR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
350383 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7Z601 | |
GPR142 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |