missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Llama IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™

Artikelnummer. 18524944 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.5 mg
Packungsgröße:
0.50 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18524944 0.5 mg 0.50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18524944 Lieferant Novus Biologicals Lieferanten-Nr. NBP175085

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Goat Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Llama samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen IgG (H+L)
Anwendungen Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konzentration 1.5 mg/mL
Konjugat Alkaline Phosphatase (AP)
Verdünnung Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin
Zusammensetzung 30 mM Triethanolamine, pH 7.2, 5 mM Magnesium Chloride, 0.1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free
Wirtsspezies Goat
Immunogen This Goat anti-Llama IgG (H+L) Secondary Antibody [Alkaline Phosphatase] was developed against purified llama IgG (H&L).
Reinigungsverfahren Affinity purified
Menge 0.5 mg
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, Immunology
Primär oder sekundär Secondary
Zielspezies Llama
Inhalt und Lagerung Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.