missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Human IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™

Artikelnummer. 18441923 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
1 mg
Packungsgröße:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18441923 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18441923 Lieferant Novus Biologicals Lieferanten-Nr. NBP172570

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Goat Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen IgG (H+L)
Anwendungen Western Blot, ELISA, Immunohistochemistry, Dot Blot
Klassifikation Polyclonal
Konjugat Alkaline Phosphatase (AP)
Verdünnung Western Blot 1:500 - 1:2500, ELISA 1:5000, Immunohistochemistry 1:200 - 1:1000, Dot Blot
Zusammensetzung 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Wirtsspezies Goat
Immunogen Human IgG whole molecule
Reinigungsverfahren Immunogen affinity purified
Menge 1 mg
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, Immunology
Primär oder sekundär Secondary
Zielspezies Human
Inhalt und Lagerung Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.