missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Feline IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™

Artikelnummer. 18445633 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
1 mg
Packungsgröße:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18445633 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18445633 Lieferant Novus Biologicals Lieferanten-Nr. NBP172587

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Goat Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Anwendungen Western Blot, ELISA, Immunohistochemistry
Klassifikation Polyclonal
Konjugat Alkaline Phosphatase (AP)
Verdünnung Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:6000, Immunohistochemistry 1:200 - 1:1000
Zusammensetzung 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Wirtsspezies Goat
Immunogen Cat IgG whole molecule
Reinigungsverfahren Multi-step
Menge 1 mg
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, Immunology
Primär oder sekundär Secondary
Zielspezies Feline
Inhalt und Lagerung Store at 4°C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.