missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNGT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11884-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Anti-GNGT1 (11884-1-AP) is produced by immunizing rabbits with full-length chain of GNGT1. The antibody detects an 8 kDa band in mouse retinal tissue.
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein- effector interaction.Spezifikation
GNGT1 | |
Polyclonal | |
Unconjugated | |
GNGT1 | |
GNG1, GNGT1, Transducin gamma chain | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14699, 2792, 680149 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P63211, Q61012 | |
GNGT1 | |
GNGT1 Fusion Protein Ag2488 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |