missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNB4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | GNB4 |
---|---|
Konzentration | 0.15 mg/mL |
Anwendungen | Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839413
|
Proteintech
11978-2-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829413
|
Proteintech
11978-2-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors.Spezifikation
GNB4 | |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O35353, P29387, Q9HAV0 | |
14696, 294962, 59345 | |
GNB4 Fusion Protein Ag2604 | |
Primary | |
-20°C | |
GNB4 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GNB4, Transducin beta chain 4 | |
GNB4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |